missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254 come in a variety of sizes.
27.20 € - 271.00 €
Specifications
Particle Size | 5 to 17 μm |
---|---|
Pore Size | 60 Å |
Fluorescent Indicator | UV254 |
Pore Volume | 0.75 mL/g |
Specific Surface | ∼500 m2/g |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
12820951
|
Macherey-Nagel™
818129 |
2.5 x 7.5 cm |
175.00 €
Pack of 200 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
10517771
|
Macherey-Nagel™
818133 |
20 x 20 cm |
271.00 €
Pack of 25 |
In Stock
Log in to see stock. |
|||||
10451193
|
Macherey-Nagel™
818131 |
4 x 8 cm |
41.05 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
12860651
|
Macherey-Nagel™
818132 |
5 x 20 cm |
77.70 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
10115001
|
Macherey-Nagel™
818130.20 |
5 x 7.5 cm |
27.20 €
Pack of 20 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
10756342
|
Macherey-Nagel™
818160 |
5 x 10 cm |
77.40 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
12834031
|
Macherey-Nagel™
818162 |
10 x 20 cm |
72.15 €
Pack of 20 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
Description
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254 come in a variety of sizes.
Alugram™ Aluminum Sheets for TLC- Pre-coated ready to use plates
- Broad range of plate sizes, surface coatings and backings
- Silica 60
- specific surface (BET) ∼ 500 m2/g
- Mean pore size 60 Å
- Specific pore volume 0.75 mL/g,
- Particle size 5–17 μm
- Standard grade
- For analytical plates 0.25 mm
- For preparative plates 0.5 and 1 mm
- For 2 mm preparative layers a slightly coarser material is used.
- Manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
- Special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm)
- Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents
Specifications
5 to 17 μm | |
UV254 | |
∼500 m2/g |
60 Å | |
0.75 mL/g |