missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Chemical Oxygen Demand (COD) Test Kits

Product Code. 11717315
Change view
Click to view available options
No. of Tests:
25
Unit Size:
0.1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests unitSize
11717315 25 0.1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11717315 Supplier WTW™ Supplier No. 252070

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Obtain precision measurements for wastewater, production control, ground water, surface water with the Chemical Oxygen Demand (COD) test kits.

  • For wastewater, production control, ground water, surface water
  • For use with PhotoLab™6, PhotoLab™ S12, PhotoLab™ 6000, PhotoLab™ Spektral photometer, Photoflex™ meter
  • Chemical Oxygen Demand (COD) Test Kits
  • This test is highly accurate and easy to use
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN ISO 15705
Volume (Metric) 3 mL
For Use With (Application) Wastewater, production control, ground water, surface water
For Use With (Equipment) PhotoLab™6, PhotoLab™ S12, PhotoLab™ 6000, PhotoLab™ Spektral photometer, Ph
No. of Tests 25
compliance-icons compliance-icons compliance-icons
Product Identifier
  • COD Cell Test C3/25 Method: photometric 10 - 150 mg/l
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, mercury(II) sulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.