missing translation for 'onlineSavingsMsg'
Learn More

Ethyl vinyl ketone, 97%, stab.

Product Code. p-7044501
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11386996

Brand: Thermo Scientific Alfa Aesar L06829.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1629-58-9 | C5H8O | 84.118 g/mol

Ethyl vinyl ketone is used as reagent for annulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl vinyl ketone is used as reagent for annulation.

Solubility
Insoluble in water; soluble in most organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1629-58-9
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00009316
InChI Key JLIDVCMBCGBIEY-UHFFFAOYSA-N
Synonym 1-penten-3-one, ethyl vinyl ketone, ethylvinylketone, 1-pentene-3-one, ethylvinyl ketone, pentenone, ketone, ethyl vinyl, vinyl ethyl ketone, unii-r0053y1az7, fema no. 3382
PubChem CID 15394
IUPAC Name pent-1-en-3-one
SMILES CCC(=O)C=C

Specifications

Density 0.844
Boiling Point 103°C to 104°C
Flash Point −6°C (21°F)
Odor Pungent
Refractive Index 1.42
Quantity 5 g
UN Number UN3286
Beilstein 1735857
Solubility Information Insoluble in water; soluble in most organic solvents.
Formula Weight 84.12
Percent Purity 97%
Chemical Name or Material Ethyl vinyl ketone, stabilized
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.