Learn More
Fisherbrand™ Traceable™ Refrigerator/Freezer Alarm Thermometers
Provides accurate reading when freezer/refrigerator doors are open with temperature-buffered, sensor-sealed in bottled glycol solution
Marque: Fisherbrand™
Code : 50
Informations supplémentaires : Weight : 0.18000kg
Includes: Velcro™ and magnetic mounting strips, one AAA battery, and ABS plastic case
Description
Provides accurate reading when freezer/refrigerator doors are open with temperature-buffered, sensor-sealed in bottled glycol solution. Fisher Scientific™ Traceable™ Refrigerator/Freezer Alarm Thermometer shows current, high and low temperatures and alerts user with visual and audible signals if temperature diverts from set points.
- Monitors temperature in freezers, refrigerators, incubators, water baths and heating blocks
- Triple display simultaneously shows current, high and low temperatures
- Alarm (Cat. No. 06-664-11 only) set in 1° increments alerts user with visual and audible signals if temperature goes outside setpoints
- 10 ft. long (3m) micro cable permits doors to close on it
- Dimensions: 4.25L × 2.75W × 0.75 in.H (11 × 7 × 2cm)
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.
Spécification
ISO 17025, A2LA, NIST | |
Monitoring temperature in freezers, refrigerators, incubators, heating blocks | |
16 mm | |
2.5 in. | |
63.5 mm | |
4 oz. | |
3.25 in. | |
158°F | |
-58°F | |
-58°F to +158°F | |
Yes | |
±0.5°C | |
82.5 mm |
2.5 x 3.25 x 0.63 in. (63.5 x 82.5 x 16 mm) | |
0.63 in. | |
Velcro™ and magnetic mounting strip, one AAA battery, ABS Plastic case | |
63.5 mm | |
0.1° | |
0.11 kg | |
Bottle Probe | |
70°C | |
-50°C | |
-50°C to +70°C | |
Monitoring Temperature in Freezer, Refrigerator, Incubator, Heating Block | |
Digital LCD |