missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Fischer Aqualine™ Matrix K, for Karl Fischer Titration By Volumetry, Fisher Chemical™
Shop All Fisher Chemical ProductsDescription
- Matrix to be used in conjunction with Complete 5K (AL2250R-1) for high water content analysis of aldehydes and ketones
Specifications
Specifications
| Chemical Name or Material | Aqualine™ Matrix K |
| Name Note | Karl Fischer Reagent |
| Color | Colorless |
| CAS | 111-76-2, 107-21-1, 5683-33-0 |
| Physical Form | Liquid |
| Quantity | 1 L |
| MDL Number | 81734 |
| Packaging | Amber glass bottle |
Safety and Handling
Product Identifier
- Aqualine Matrix-K
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- H226-Flammable liquid and vapour.
- H372-Causes damage to organs.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H331-Toxic if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contain: Chloroform, 2-Butoxyethanol, Ethyl alcohol, Ethylene glycol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?