CAS RN 101990-45-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Bromo-5-(bromomethyl)pyridine, TRC
CAS: 101990-45-8 Molecular Formula: C6 H5 Br2 N Molecular Weight (g/mol): 250.919 Synonym: 6-Bromo-3-pyridylmethyl Bromide IUPAC Name: 2-bromo-5-(bromomethyl)pyridine SMILES: BrCc1ccc(Br)nc1