CAS RN 10318-16-8
CAS RN 10318-16-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Chloramphenicol 3-Acetate, TRC
CAS: 10318-16-8 Molecular Formula: C13H14Cl2N2O6 Molecular Weight (g/mol): 365.17 Synonym: 3-Acetoxychloramphenicol,3-Acetylchloramphenicol,3-O-Monoacetylchloramphenicol,[R-(R*,R*)]-N-[1-[(acetyloxy)methyl]-2-hydroxy-2-(4-nitrophenyl)ethyl]-2,2-dichloroacetamide,D-threo-(-)-2,2-Dichloro-N-[beta-hydroxy-alpha-(hydroxymethyl)-p-nitrophenethyl]acetamide alpha-Acetate IUPAC Name: [(2R,3R)-2-[(2,2-dichloroacetyl)amino]-3-hydroxy-3-(4-nitrophenyl)propyl] acetate SMILES: CC(=O)OC[C@@H](NC(=O)C(Cl)Cl)[C@H](O)c1ccc(cc1)[N+](=O)[O-]