accessibility menu, dialog, popup

UserName

CAS RN 10359-64-5

IUPAC Name: 2-(methylsulfanyl)butane
Synonyms: 2-(methylsulfanyl)butane
Molecular Weight (g/mol): 104.21
Molecular Formula: C5H12S
InChi Key: IJRCRFQMYAJPPO-UHFFFAOYNA-N
SMILES: CCC(C)SC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results