CAS RN 1041424-77-4
CAS RN 1041424-77-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
[6-(Trimethylammonium)hexyl] Methanethiosulfonate Bromide, TRC
CAS: 1041424-77-4 Molecular Formula: C10 H24 N O2 S2 . Br Molecular Weight (g/mol): 334.34 IUPAC Name: trimethyl(6-methylsulfonylsulfanylhexyl)azanium;bromide SMILES: [Br-].C[N+](C)(C)CCCCCCSS(=O)(=O)C