accessibility menu, dialog, popup

UserName

Ge

CAS RN 1048-05-1

IUPAC Name: tetraphenylgermane
Synonyms: tetraphenylgermane
Molecular Weight (g/mol): 381.05
Molecular Formula: C24H20Ge
InChi Key: ILEXMONMGUVLRM-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)[Ge](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results