CAS RN 10485-23-1
CAS RN 10485-23-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Ethoxy-N,N,N-trimethyl-2,4-dioxo-1-butanaminium Chloride, TRC
CAS: 10485-23-1 Molecular Formula: C9H18NO3 • Cl Molecular Weight (g/mol): 223.7 Synonym: (3-Carboxyacetonyl)trimethylammonium Chloride Ethyl Ester,(3-Carboxy-2-hydroxyallyl)trimethyl-ammonium Chloride Ethyl Ester IUPAC Name: 4-ethoxy-N,N,N-trimethyl-2,4-dioxobutan-1-aminium chloride SMILES: [Cl-].O=C(OCC)CC(=O)C[N+](C)(C)C