accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 10519-20-7

IUPAC Name: ethyl oct-2-ynoate
Synonyms: ethyl oct-2-ynoate
Molecular Weight (g/mol): 168.24
Molecular Formula: C10H16O2
InChi Key: QPMDWYXUSMRVKT-UHFFFAOYSA-N
SMILES: CCCCCC#CC(=O)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results