accessibility menu, dialog, popup

UserName

HBrH3CNH3CBr

CAS RN 1069-72-3

IUPAC Name: hydrogen (2-bromoethyl)diethylamine bromide
Synonyms: hydrogen (2-bromoethyl)diethylamine bromide
Molecular Weight (g/mol): 261
Molecular Formula: C6H15Br2N
InChi Key: HLMHCDKXKXBKQK-UHFFFAOYSA-N
SMILES: [H+].[Br-].CCN(CC)CCBr
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results