accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNHNOOH3CCH3H3C

CAS RN 1072944-99-0

IUPAC Name: tert-butyl N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
Synonyms: tert-butyl N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
Molecular Weight (g/mol): 320.2
Molecular Formula: C16H25BN2O4
InChi Key: FYKIZSWVZIHFLF-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=C(C=CC=N1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results