CAS RN 1073-11-6
CAS RN 1073-11-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Methyl-4-vinyl-4-butanolide, TRC
CAS: 1073-11-6 Molecular Formula: C7 H10 O2 Molecular Weight (g/mol): 126.15 Synonym: 4-Methyl-4-vinylbutyrolactone,4-Methyl-5-hexen-4-olide,5-Methyl-5-vinyldihydrofuran-2(3H)-one,Dihydro-5-methyl-5-vinyl-2(3H)-furanone,Lavender Lactone,gamma-Methyl-gamma-vinyl-gamma-butyrolactone,gamma-Vinyl-gamma-valerolactone,5-Ethenyldihydro-5-methyl-2(3H)-furanone,2-Vinyl-2-methyltetrahydrofuran-5-one,4-Methyl-4-hydroxy-5-hexenoic Acid Lactone IUPAC Name: 5-ethenyl-5-methyloxolan-2-one SMILES: CC1(CCC(=O)O1)C=C