CAS RN 1073556-31-6
CAS RN 1073556-31-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-(-)-2-(Trifluoromethyl)pyrrolidine, 95%, Thermo Scientific™
CAS: 1073556-31-6 Molecular Formula: C5H8F3N Molecular Weight (g/mol): 139.121 MDL Number: MFCD07784180 InChI Key: VINAMCOZNJHNIH-SCSAIBSYSA-N Synonym: r---2-trifluoromethyl pyrrolidine,r-2-trifluoromethyl pyrrolidine,r-2-trifluoromethylpyrrolidine,2 r-2-trifluoromethylpyrrolidine,2r-2-trifluoromethyl pyrrolidine,r-2-trifluoromethyl-pyrrolidine,2-r-2-trifluoromethylpyrrolidine,2 r-2-trifluoromethyl pyrrolidine,2r-2-trifluoromethyl-pyrrolidine PubChem CID: 7015082 IUPAC Name: (2R)-2-(trifluoromethyl)pyrrolidine SMILES: C1CC(NC1)C(F)(F)F