CAS RN 107410-07-1
CAS RN 107410-07-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Methoxybiphenyl-3-carboxylic Acid, TRC
CAS: 107410-07-1 Molecular Formula: C14H12O3 Molecular Weight (g/mol): 228.24 Synonym: 4-Methoxy[1,1'-biphenyl]-3-carboxylic Acid,2-Methoxy-5-phenylbenzoic Acid IUPAC Name: 2-methoxy-5-phenylbenzoic acid SMILES: COc1ccc(cc1C(=O)O)c2ccccc2