accessibility menu, dialog, popup

UserName

CAS RN 1080-43-9

IUPAC Name: dimethyldiphenylstannane
Synonyms: dimethyldiphenylstannane
Molecular Weight (g/mol): 302.99
Molecular Formula: C14H16Sn
InChi Key: INCQQSKGFIBXAY-UHFFFAOYSA-N
SMILES: C[Sn](C)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results