accessibility menu, dialog, popup

UserName

CAS RN 1086138-36-4

IUPAC Name: bis({2-[bis(adamantan-1-yl)phosphanyl]ethyl})amine
Synonyms: bis({2-[bis(adamantan-1-yl)phosphanyl]ethyl})amine
Molecular Weight (g/mol): 673.99
Molecular Formula: C44H69NP2
InChi Key: KBHQOEPPHGCYHC-UHFFFAOYSA-N
SMILES: C(CP(C12CC3CC(CC(C3)C1)C2)C12CC3CC(CC(C3)C1)C2)NCCP(C12CC3CC(CC(C3)C1)C2)C12CC3CC(CC(C3)C1)C2
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results