accessibility menu, dialog, popup

UserName

CH3N

CAS RN 109-06-8

IUPAC Name: 2-methylpyridine
Synonyms: α-methylpyridine rcra waste number U191 2-mepy
Molecular Weight (g/mol): 93.13
Molecular Formula: C6H7N
InChi Key: BSKHPKMHTQYZBB-UHFFFAOYSA-N
SMILES: CC1=CC=CC=N1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)