accessibility menu, dialog, popup

UserName

CAS RN 1092461-21-6

ClOHOOFFF

CAS RN 1092461-21-6

IUPAC Name: 2-[5-chloro-2-(trifluoromethoxy)phenyl]acetic acid
Synonyms: [5-chloro-2-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol): 254.59
Molecular Formula: C9H6ClF3O3
InChi Key: KXUCNLJTXUDMHA-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(OC(F)(F)F)C=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results