CAS RN 1092978-76-1
CAS RN 1092978-76-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Diethylaminoethanol-d10 Hydrochloride, TRC
CAS: 1092978-76-1 Molecular Formula: C6H6D10ClNO Molecular Weight (g/mol): 163.71 Synonym: (Diethylamino)ethanol-d10 Hydrochloride,Dehydasal-d10 Hydrochloride,Perdilaton-d10 Hydrochloride SMILES: Cl.[2H]C([2H])([2H])C([2H])([2H])N(CCO)C([2H])([2H])C([2H])([2H])[2H]