accessibility menu, dialog, popup

UserName

CAS RN 110-53-2

IUPAC Name: 1-bromopentane
Synonyms: 1-bromopentane
Molecular Weight (g/mol): 151.05
Molecular Formula: C5H11Br
InChi Key: YZWKKMVJZFACSU-UHFFFAOYSA-N
SMILES: CCCCCBr
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)