accessibility menu, dialog, popup

UserName

CAS RN 110-65-6

OHHO

CAS RN 110-65-6

IUPAC Name: but-2-yne-1,4-diol
Synonyms: butynediol 1,4-butinodiol agrisynth B3D 2-butin-1,4-diol 1,4-dimethoxyacetylene
Molecular Weight (g/mol): 86.09
Molecular Formula: C4H6O2
InChi Key: DLDJFQGPPSQZKI-UHFFFAOYSA-N
SMILES: OCC#CCO
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)