accessibility menu, dialog, popup

UserName

O

CAS RN 110-87-2

IUPAC Name: 3,4-dihydro-2H-pyran
Synonyms: dihydropyran
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: BUDQDWGNQVEFAC-UHFFFAOYSA-N
SMILES: C1COC=CC1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)