accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 111-12-6

IUPAC Name: methyl oct-2-ynoate
Synonyms: methyl 2-octynoate methyl heptine carbonate
Molecular Weight (g/mol): 154.21
Molecular Formula: C9H14O2
InChi Key: FRLZQXRXIKQFNS-UHFFFAOYSA-N
SMILES: CCCCCC#CC(=O)OC
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results