accessibility menu, dialog, popup

UserName

H3COOOCH3O

CAS RN 111-55-7

IUPAC Name: 2-(acetyloxy)ethyl acetate
Synonyms: ethylene glycol diacetate
Molecular Weight (g/mol): 146.14
Molecular Formula: C6H10O4
InChi Key: JTXMVXSTHSMVQF-UHFFFAOYSA-N
SMILES: CC(=O)OCCOC(C)=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (6)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (3)
  • (3)
  • (6)