accessibility menu, dialog, popup

UserName

CH3NH2ONH2O

CAS RN 1113-63-9

IUPAC Name: 2-methylpropanediamide
Synonyms: 2-methylpropanediamide
Molecular Weight (g/mol): 116.12
Molecular Formula: C4H8N2O2
InChi Key: CGRKYGKHZOCPSZ-UHFFFAOYSA-N
SMILES: CC(C(N)=O)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results