CAS RN 1117-41-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methylisopseudoionone, TRC
CAS: 1117-41-5 Molecular Formula: C14 H22 O Molecular Weight (g/mol): 206.32 Synonym: Pseudoionone, isomethyl- (5CI),3,6,10-Trimethyl-3,5,9-undecatrien-2-one,Isomethylpseudoionone,Methylisopseudoionone IUPAC Name: (3E,5E)-3,6,10-trimethylundeca-3,5,9-trien-2-one SMILES: CC(=CCC\C(=C\C=C(/C)\C(=O)C)\C)C