accessibility menu, dialog, popup

UserName

NH2NNN

CAS RN 1120-99-6

IUPAC Name: 1,2,4-triazin-3-amine
Synonyms: 3-amino-1,2,4-triazine
Molecular Weight (g/mol): 96.09
Molecular Formula: C3H4N4
InChi Key: MJIWQHRXSLOUJN-UHFFFAOYSA-N
SMILES: NC1=NC=CN=N1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)