CAS RN 112725-22-1
CAS RN 112725-22-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Succinimidyl 3-(Tri-n-butylstannyl)benzoate, TRC
CAS: 112725-22-1 Molecular Formula: C23H35NSnO4 Molecular Weight (g/mol): 508.24 Synonym: 3-(tributylstannyl)-Benzoic acid 2,5-dioxo-1-pyrrolidinyl ester,1-[[3-(tributylstannyl)benzoyl]oxy]-2,5-Pyrrolidinedione (9CI) IUPAC Name: (2,5-dioxopyrrolidin-1-yl) 3-tributylstannylbenzoate SMILES: CCCC[Sn](CCCC)(CCCC)c1cccc(c1)C(=O)ON2C(=O)CCC2=O