CAS RN 112859-71-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fluasterone, TRC
CAS: 112859-71-9 Molecular Formula: C19 H27 F O Molecular Weight (g/mol): 290.42 Synonym: (16α)-Fluoro-5-androsten-17-one; IUPAC Name: (8R,9S,10R,13S,14S,16R)-16-fluoro-10,13-dimethyl-1,2,3,4,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-one SMILES: C[C@]12CC[C@H]3[C@@H](CC=C4CCCC[C@]34C)[C@@H]1C[C@@H](F)C2=O