accessibility menu, dialog, popup

UserName

ClN

CAS RN 112905-95-0

IUPAC Name: 1-(chloromethyl)cyclohexane-1-carbonitrile
Synonyms: 1-(chloromethyl)cyclohexane-1-carbonitrile
Molecular Weight (g/mol): 157.64
Molecular Formula: C8H12ClN
InChi Key: BWQNUOFYAYIAIY-UHFFFAOYSA-N
SMILES: ClCC1(CCCCC1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results