CAS RN 113931-96-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methoxy4-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-3-benzenemethanol, TRC
CAS: 113931-96-7 Molecular Formula: C14H24O3Si Molecular Weight (g/mol): 268.43 Synonym: 4-tert-Butyldimethylsilyloxy-3-methoxybenzyl Alcohol IUPAC Name: [4-[tert-butyl(dimethyl)silyl]oxy-3-methoxyphenyl]methanol SMILES: COc1cc(CO)ccc1O[Si](C)(C)C(C)(C)C