accessibility menu, dialog, popup

UserName

OOSOOCl

CAS RN 115010-10-1

IUPAC Name: 2H-1,3-benzodioxole-5-sulfonyl chloride
Synonyms: 2H-1,3-benzodioxole-5-sulfonyl chloride
Molecular Weight (g/mol): 220.62
Molecular Formula: C7H5ClO4S
InChi Key: ICUBASIDCXDQAW-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC2=C(OCO2)C=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results