CAS RN 115016-95-0
IUPAC Name:
2-hydroxy-4-phenylbutanoic acid
Synonyms:
2-hydroxy-4-phenylbutanoic acid
Molecular Weight (g/mol):
180.2
Molecular Formula:
C10H12O3
InChi Key:
JNJCEALGCZSIGB-UHFFFAOYNA-N
SMILES:
OC(CCC1=CC=CC=C1)C(O)=O