accessibility menu, dialog, popup

UserName

BFFFFFFFFFK

CAS RN 1150655-10-9

IUPAC Name: potassium [2,4-bis(trifluoromethyl)phenyl]trifluoroboranuide
Synonyms: potassium [2,4-bis(trifluoromethyl)phenyl]trifluoroboranuide
Molecular Weight (g/mol): 320.01
Molecular Formula: C8H3BF9K
InChi Key: OXVQSBLFWHNZJA-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=C(C=C(C=C1)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results