accessibility menu, dialog, popup

UserName

CAS RN 116344-17-3

IUPAC Name: 1-phenyl-5-propyl-1H-pyrazole-4-carboxylic acid
Synonyms: 1-phenyl-5-propylpyrazole-4-carboxylic acid
Molecular Weight (g/mol): 230.27
Molecular Formula: C13H14N2O2
InChi Key: CBYITWPSSDTYTN-UHFFFAOYSA-N
SMILES: CCCC1=C(C=NN1C1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results