accessibility menu, dialog, popup

UserName

CAS RN 116525-66-7

H3CSSOOHClN

CAS RN 116525-66-7

IUPAC Name: 3-(4-chlorophenyl)-4-cyano-5-(methylsulfanyl)thiophene-2-carboxylic acid
Synonyms: 3-(4-chlorophenyl)-4-cyano-5-(methylsulfanyl)thiophene-2-carboxylic acid
Molecular Weight (g/mol): 309.78
Molecular Formula: C13H8ClNO2S2
InChi Key: CCQQTQYIJRVXIU-UHFFFAOYSA-N
SMILES: CSC1=C(C#N)C(=C(S1)C(O)=O)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results