accessibility menu, dialog, popup

UserName

ClOOO

CAS RN 117-21-5

IUPAC Name: 4-chloro-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 3-chlorophthalic anhydride
Molecular Weight (g/mol): 182.56
Molecular Formula: C8H3ClO3
InChi Key: UERPUZBSSSAZJE-UHFFFAOYSA-N
SMILES: ClC1=CC=CC2=C1C(=O)OC2=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)