CAS RN 117078-70-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-[(4-Chlorophenyl)methyl]-2-[2-(1-methyl-2-pyrrolidinyl)ethyl]-1(2H)-phthalazinone Hydrochloride, TRC
CAS: 117078-70-3 Molecular Formula: C22H24ClN3O . HCl Molecular Weight (g/mol): 381.9 Synonym: Azelastine 5-Member-Cyclic-Isomer Hcl IUPAC Name: 4-[(4-chlorophenyl)methyl]-2-[2-(1-methylpyrrolidin-2-yl)ethyl]phthalazin-1-one SMILES: Cl.CN1CCCC1CCN2N=C(Cc3ccc(Cl)cc3)c4ccccc4C2=O