CAS RN 117977-18-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(3-Methoxypropoxy)-2,3-dimethylpyridin-1-ium-1-olate, TRC
CAS: 117977-18-1 Molecular Formula: C11H17NO3 Molecular Weight (g/mol): 211.25 Synonym: 4-(3-Methoxypropoxy)-2,3-dimethylpyridine N-Oxide IUPAC Name: 4-(3-methoxypropoxy)-2,3-dimethyl-1-oxidopyridin-1-ium SMILES: COCCCOc1cc[n+]([O-])c(C)c1C