CAS RN 118054-54-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1-Methyl-1H-imidazol-2-yl)-acetic Acid, TRC
CAS: 118054-54-9 Molecular Formula: C6H8N2O2 Molecular Weight (g/mol): 140.14 IUPAC Name: 2-(1-methylimidazol-2-yl)acetic acid SMILES: Cn1ccnc1CC(=O)O