accessibility menu, dialog, popup

UserName

H3COBOHHOOH3COCH3

CAS RN 118062-05-8

IUPAC Name: (2,3,4-trimethoxyphenyl)boronic acid
Synonyms: 2,3,4-trimethoxyphenylboronic acid
Molecular Weight (g/mol): 212.01
Molecular Formula: C9H13BO5
InChi Key: LDQLSQRFSNMANA-UHFFFAOYSA-N
SMILES: COC1=CC=C(B(O)O)C(OC)=C1OC
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results