CAS RN 1182-42-9
CAS RN 1182-42-9
IUPAC Name:
(1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl octanoate
Synonyms:
cholesterol n-octanoate
Molecular Weight (g/mol):
512.86
Molecular Formula:
C35H60O2
InChi Key:
SKLBBRQPVZDTNM-SJTWHRLHSA-N
SMILES:
CCCCCCCC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCCC(C)C
1
–
1
of
1
results
1
Cholesterol Caprylate, TRC
CAS: 1182-42-9 Molecular Formula: C35 H60 O2 Molecular Weight (g/mol): 512.85 Synonym: (3β)-Cholest-5-en-3-ol Octanoate,Cholesterol Octanoate,3β-(Octanoyloxy)cholest-5-ene,5-Cholesten-3β-ol Caprylate,Caprylic Acid Cholesterol Ester,Cholest-5-en-3β-ol caprylate,Cholesteryl Caprylate,Cholesteryl Octanoate IUPAC Name: [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] octanoate SMILES: CCCCCCCC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCCC(C)C