accessibility menu, dialog, popup

UserName

CAS RN 118276-06-5

FHOFO

CAS RN 118276-06-5

IUPAC Name: 3,5-difluoro-4-hydroxybenzaldehyde
Synonyms: 3,5-difluoro-4-hydroxybenzaldehyde
Molecular Weight (g/mol): 158.1
Molecular Formula: C7H4F2O2
InChi Key: SKOYTQILPMNZQO-UHFFFAOYSA-N
SMILES: OC1=C(F)C=C(C=O)C=C1F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results