CAS RN 1184-57-2
IUPAC Name:
methylmercurylium hydroxide
Synonyms:
methyl mercury ion hydroxide
Molecular Weight (g/mol):
232.63
Molecular Formula:
CH4HgO
InChi Key:
KRZWEBVPFGCYMY-UHFFFAOYSA-M
SMILES:
[OH-].C[Hg+]
1
–
2
of
2
results
1
Methylmercury(II) hydroxide, 1M in H{2}O
CAS: 1184-57-2 MDL Number: MFCD00013924
2
Methylmercury(II) hydroxide, 1M in H2O, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™
CAS: 1184-57-2 MDL Number: MFCD00013924