accessibility menu, dialog, popup

UserName

CAS RN 118486-94-5

IUPAC Name: tributyl(furan-2-yl)stannane
Synonyms: tributyl(furan-2-yl)stannane
Molecular Weight (g/mol): 357.13
Molecular Formula: C16H30OSn
InChi Key: SANWDQJIWZEKOD-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=CC=CO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results