accessibility menu, dialog, popup

UserName

CH3GeHH3CH3C

CAS RN 1188-14-3

IUPAC Name: triethylgermane
Synonyms: triethylgermane
Molecular Weight (g/mol): 160.82
Molecular Formula: C6H16Ge
InChi Key: LTLGQZPDSQYFSB-UHFFFAOYSA-N
SMILES: CC[GeH](CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results