CAS RN 1189729-40-5
CAS RN 1189729-40-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Losartan-d3 Carboxylic Acid, TRC
CAS: 1189729-40-5 Molecular Formula: C22 2H3 H18 Cl N6 O2 Molecular Weight (g/mol): 439.91 IUPAC Name: 5-chloro-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]-2-(4,4,4-trideuteriobutyl)imidazole-4-carboxylic acid SMILES: [2H]C([2H])([2H])CCCc1nc(Cl)c(C(=O)O)n1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4